Assay Details
Gene Symbol: abr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100016.1 | NP_001093486.1 | 7 - 8 | 881 | 69 | |
NM_001368694.1 | NP_001355623.1 | 22 - 23 | 3378 | 69 | ||
XM_003198714.5 | XP_003198762.4 | 22 - 23 | 2760 | 69 | ||
XM_005168629.4 | XP_005168686.1 | 7 - 8 | 1185 | 69 | ||
XM_005172023.4 | XP_005172080.3 | 22 - 23 | 2443 | 69 | ||
XM_017356374.2 | XP_017211863.2 | 17 - 18 | 1884 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
