Assay Details
Gene Symbol: trioa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104526.1 | NP_001097996.1 | 26 - 27 | 4165 | 73 | |
XM_009294421.3 | XP_009292696.1 | 26 - 27 | 4332 | 73 | ||
XM_017352243.2 | XP_017207732.1 | 26 - 27 | 4336 | 73 | ||
XM_021468075.1 | XP_021323750.1 | 26 - 27 | 4395 | 73 | ||
XM_021468076.1 | XP_021323751.1 | 26 - 27 | 4396 | 73 | ||
XM_021468077.1 | XP_021323752.1 | 26 - 27 | 4396 | 73 | ||
XM_021468078.1 | XP_021323753.1 | 26 - 27 | 4397 | 73 | ||
XM_021468079.1 | XP_021323754.1 | 26 - 27 | 4398 | 73 | ||
XM_021468080.1 | XP_021323755.1 | 26 - 27 | 4399 | 73 | ||
XM_021468081.1 | XP_021323756.1 | 26 - 27 | 4399 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
