Assay Details
Gene Symbol: guca1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131870.1 | NP_571945.1 | 2 - 3 | 418 | 80 | |
GenBank mRNA | AY044455.1 | - | 2 - 3 | 416 | 80 | |
AY050501.1 | - | 2 - 3 | 418 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
