Assay Details
Gene Symbol: guca1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131871.1 | NP_571946.1 | 1 - 2 | 426 | 87 | |
GenBank mRNA | AY044456.1 | - | 1 - 2 | 423 | 87 | |
AY050502.1 | - | 1 - 2 | 426 | 87 | ||
BC124761.1 | - | 1 - 2 | 422 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
