Assay Details
Gene Symbol: guca1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131871.1 | NP_571946.1 | 3 - 4 | 691 | 73 | |
GenBank mRNA | AY044456.1 | - | 3 - 4 | 688 | 73 | |
AY050502.1 | - | 3 - 4 | 691 | 73 | ||
BC124761.1 | - | 3 - 4 | 687 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
