Assay Details
Gene Symbol: mkrn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_152979.2 | NP_694511.1 | 4 - 5 | 750 | 66 | |
XM_005155668.4 | XP_005155725.1 | 4 - 5 | 768 | 66 | ||
GenBank mRNA | AF277172.1 | - | 4 - 5 | 699 | 66 | |
BC045986.1 | - | 4 - 5 | 66 | |||
BC065352.1 | - | 4 - 5 | 694 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
