Assay Details
Gene Symbol: sdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173223.2 | NP_775330.2 | 3 - 4 | 601 | 101 | |
GenBank mRNA | AF262048.1 | - | 3 - 4 | 398 | 101 | |
AY091914.1 | - | 3 - 4 | 338 | 101 | ||
BC081509.1 | - | 3 - 4 | 601 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
