Assay Details
Gene Symbol: gria3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198360.2 | NP_938174.1 | 11 - 12 | 1901 | 124 | |
XM_005157132.3 | XP_005157189.1 | 11 - 12 | 2036 | 124 | ||
XM_005157133.3 | XP_005157190.1 | 11 - 12 | 2035 | 124 | ||
XM_009307719.3 | XP_009305994.1 | 11 - 12 | 2047 | 124 | ||
XM_021480987.1 | XP_021336662.1 | 11 - 12 | 2049 | 124 | ||
GenBank mRNA | AF525746.1 | - | 11 - 12 | 1901 | 124 | |
EB928785.1 | - | 1 - 2 | 387 | 124 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
