Assay Details
Gene Symbol: gria3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198360.2 | NP_938174.1 | 13 - 14 | 2342 | 100 | |
XM_005157132.3 | XP_005157189.1 | 13 - 14 | 2477 | 100 | ||
XM_021480987.1 | XP_021336662.1 | 13 - 14 | 2490 | 100 | ||
GenBank mRNA | AF525746.1 | - | 13 - 14 | 2342 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
