Assay Details
Gene Symbol: gria3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198360.2 | NP_938174.1 | 2 - 3 | 293 | 113 | |
XM_005157132.3 | XP_005157189.1 | 2 - 3 | 428 | 113 | ||
XM_005157133.3 | XP_005157190.1 | 2 - 3 | 427 | 113 | ||
XM_009307719.3 | XP_009305994.1 | 2 - 3 | 439 | 113 | ||
XM_021480987.1 | XP_021336662.1 | 2 - 3 | 441 | 113 | ||
GenBank mRNA | AF525746.1 | - | 2 - 3 | 293 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
