Assay Details
Gene Symbol: paics | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199537.2 | NP_955831.1 | 4 - 5 | 731 | 117 | |
GenBank mRNA | BC048051.1 | - | 4 - 5 | 694 | 117 | |
BC071527.1 | - | 4 - 5 | 731 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
