Assay Details
Gene Symbol: paics | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199537.2 | NP_955831.1 | 8 - 9 | 1278 | 71 | |
GenBank mRNA | BC048051.1 | - | 8 - 9 | 1241 | 71 | |
BC071527.1 | - | 8 - 9 | 1278 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
