Assay Details
Gene Symbol: msrb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212921.2 | NP_998086.2 | 3 - 4 | 552 | 83 | |
GenBank mRNA | BC067722.1 | - | 394 | 83 | ||
EH490030.1 | - | 3 - 4 | 588 | 83 | ||
EH509367.1 | - | 3 - 4 | 409 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
