Assay Details
Gene Symbol: gadd45ba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213031.3 | NP_998196.1 | 1 - 2 | 206 | 75 | |
GenBank mRNA | AB180736.1 | - | 1 - 2 | 135 | 75 | |
AY714220.1 | - | 1 - 2 | 179 | 75 | ||
BC059472.1 | - | 1 - 2 | 75 | 75 | ||
BC095628.1 | - | 1 - 2 | 209 | 75 | ||
BC164931.1 | - | 1 - 2 | 85 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
