Assay Details
Gene Symbol: adipor1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213500.1 | NP_998665.1 | 384 | 110 | ||
GenBank mRNA | BC044388.1 | - | 384 | 110 | ||
BC164776.1 | - | 1 - 2 | 303 | 110 | ||
EU139311.1 | - | 1 - 2 | 252 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
