Assay Details
Gene Symbol: cbx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205749.1 | NP_991312.1 | 5 | 1348 | 155 | |
GenBank mRNA | AY186581.1 | - | 5 | 1348 | 155 | |
BC171350.1 | - | 5 | 1435 | 155 | ||
BC171352.1 | - | 5 | 1435 | 155 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
