Assay Details
Gene Symbol: pcxb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131550.2 | NP_571625.2 | 7 | 1057 | 126 | |
GenBank mRNA | AF295372.1 | - | 7 | 850 | 126 | |
BC162583.1 | - | 7 | 850 | 126 | ||
EH613146.1 | - | 3 | 517 | 126 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
