Assay Details
Gene Symbol: c1qtnf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017875.2 | NP_001017875.1 | 4 | 2547 | 85 | |
GenBank mRNA | BC093423.1 | - | 6 | 1448 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
