Assay Details
Gene Symbol: slc17a6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005398.1 | NP_001005398.1 | 11 | 1442 | 61 | |
NM_001128821.1 | NP_001122293.1 | 12 | 1747 | 61 | ||
GenBank mRNA | AB183386.1 | - | 12 | 1749 | 61 | |
BC162508.1 | - | 12 | 1747 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
