Assay Details
Gene Symbol: acadsb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001351649.2 | NP_001338578.1 | 11 | 1649 | 93 | |
XM_021480455.1 | XP_021336130.1 | 10 | 1588 | 93 | ||
GenBank mRNA | BC135076.1 | - | 6 | 650 | 93 |
Gene Symbol: LOC100037342 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_009298405.3 | XP_009296680.1 | 8 | 1327 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
