Assay Details
Gene Symbol: hopx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198209.2 | NP_937852.1 | 2 | 149 | 93 | |
GenBank mRNA | AF536198.1 | - | 1 | 226 | 93 | |
BC096856.1 | - | 2 | 149 | 93 | ||
BC153537.1 | - | 1 | 205 | 93 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC096856.1, BC153537.1, NM_198209.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
