Assay Details
Gene Symbol: kctd5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207049.1 | NP_996932.1 | 6 | 1754 | 77 | |
GenBank mRNA | BC066418.1 | - | 6 | 1754 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
