Assay Details
Gene Symbol: sp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212662.2 | NP_997827.2 | 6 | 1916 | 148 | |
XM_017358202.1 | XP_017213691.1 | 6 | 2012 | 148 | ||
XM_021479551.1 | XP_021335226.1 | 6 | 2514 | 148 | ||
GenBank mRNA | BC067713.1 | - | 6 | 1845 | 148 |
Gene Symbol: LOC110438533 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021471073.1 | XP_021326748.1 | 3 | 1449 | 148 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
