Assay Details
Gene Symbol: dffa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002631.1 | NP_001002631.1 | 3 - 4 | 557 | 67 | |
GenBank mRNA | BC075948.1 | - | 3 - 4 | 557 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
