Assay Details
Gene Symbol: rgs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002662.2 | NP_001002662.2 | 4 - 5 | 446 | 112 | |
GenBank mRNA | BC076456.1 | - | 4 - 5 | 432 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
