Assay Details
Gene Symbol: fkbp14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003471.1 | NP_001003471.1 | 2 - 3 | 444 | 106 | |
GenBank mRNA | BC078342.1 | - | 2 - 3 | 444 | 106 | |
BC165022.1 | - | 2 - 3 | 382 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
