Assay Details
Gene Symbol: sh3bgr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003524.2 | NP_001003524.1 | 11 - 12 | 735 | 74 | |
NM_001362476.1 | NP_001349405.1 | 8 - 9 | 777 | 74 | ||
NM_001362477.1 | NP_001349406.1 | 10 - 11 | 711 | 74 | ||
NM_001362478.1 | NP_001349407.1 | 9 - 10 | 684 | 74 | ||
NM_001362479.1 | NP_001349408.1 | 7 - 8 | 579 | 74 | ||
NM_001362480.1 | NP_001349409.1 | 6 - 7 | 555 | 74 | ||
XM_005169682.4 | XP_005169739.1 | 12 - 13 | 904 | 74 | ||
XM_005169685.4 | XP_005169742.1 | 9 - 10 | 826 | 74 | ||
XM_005169686.4 | XP_005169743.1 | 11 - 12 | 789 | 74 | ||
XM_005169688.4 | XP_005169745.1 | 10 - 11 | 642 | 74 | ||
XM_009291451.3 | XP_009289726.1 | 11 - 12 | 880 | 74 | ||
XM_009291452.3 | XP_009289727.1 | 11 - 12 | 880 | 74 | ||
XM_009291453.3 | XP_009289728.1 | 10 - 11 | 856 | 74 | ||
XM_009291457.3 | XP_009289732.1 | 7 - 8 | 628 | 74 | ||
XM_021481381.1 | XP_021337056.1 | 10 - 11 | 853 | 74 | ||
XM_021481383.1 | XP_021337058.1 | 9 - 10 | 682 | 74 | ||
XM_021481384.1 | XP_021337059.1 | 9 - 10 | 679 | 74 | ||
XM_021481385.1 | XP_021337060.1 | 9 - 10 | 617 | 74 | ||
GenBank mRNA | BC077137.1 | - | 11 - 12 | 708 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
