Assay Details
Gene Symbol: sh3bgr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003524.2 | NP_001003524.1 | 2 - 3 | 367 | 82 | |
NM_001362476.1 | NP_001349405.1 | 2 - 3 | 367 | 82 | ||
NM_001362477.1 | NP_001349406.1 | 2 - 3 | 367 | 82 | ||
NM_001362478.1 | NP_001349407.1 | 2 - 3 | 367 | 82 | ||
NM_001362479.1 | NP_001349408.1 | 2 - 3 | 367 | 82 | ||
NM_001362480.1 | NP_001349409.1 | 2 - 3 | 367 | 82 | ||
XM_005169682.4 | XP_005169739.1 | 2 - 3 | 392 | 82 | ||
XM_005169685.4 | XP_005169742.1 | 2 - 3 | 392 | 82 | ||
XM_005169686.4 | XP_005169743.1 | 2 - 3 | 355 | 82 | ||
XM_005169688.4 | XP_005169745.1 | 2 - 3 | 352 | 82 | ||
XM_009291451.3 | XP_009289726.1 | 2 - 3 | 392 | 82 | ||
XM_009291452.3 | XP_009289727.1 | 2 - 3 | 392 | 82 | ||
XM_009291453.3 | XP_009289728.1 | 2 - 3 | 392 | 82 | ||
XM_009291457.3 | XP_009289732.1 | 2 - 3 | 362 | 82 | ||
XM_021481381.1 | XP_021337056.1 | 2 - 3 | 392 | 82 | ||
XM_021481383.1 | XP_021337058.1 | 2 - 3 | 362 | 82 | ||
XM_021481384.1 | XP_021337059.1 | 2 - 3 | 362 | 82 | ||
XM_021481385.1 | XP_021337060.1 | 2 - 3 | 351 | 82 | ||
GenBank mRNA | BC077137.1 | - | 2 - 3 | 340 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
