Assay Details
Gene Symbol: mphosph6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003618.1 | NP_001003618.1 | 3 - 4 | 350 | 94 | |
GenBank mRNA | BC078314.1 | - | 3 - 4 | 350 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
