Assay Details
Gene Symbol: fkbp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004506.1 | NP_001004506.1 | 1 - 2 | 215 | 61 | |
GenBank mRNA | BC163638.1 | - | 1 - 2 | 301 | 61 | |
BC163644.1 | - | 1 - 2 | 301 | 61 | ||
BC163849.1 | - | 1 - 2 | 301 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
