Assay Details
Gene Symbol: gpd1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005934.2 | NP_001005934.1 | 6 - 7 | 968 | 80 | |
GenBank mRNA | BC083522.1 | - | 6 - 7 | 948 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
