Assay Details
Gene Symbol: rtkn2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005937.2 | NP_001005937.2 | 2 - 3 | 382 | 107 | |
GenBank mRNA | BC083519.1 | - | 2 - 3 | 382 | 107 | |
BC098873.1 | - | 2 - 3 | 382 | 107 | ||
DV585005.1 | - | 1 - 2 | 217 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
