Assay Details
Gene Symbol: rtkn2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005937.2 | NP_001005937.2 | 3 - 4 | 434 | 95 | |
GenBank mRNA | BC083519.1 | - | 3 - 4 | 434 | 95 | |
BC092947.1 | - | 3 - 4 | 435 | 95 | ||
BC098873.1 | - | 3 - 4 | 434 | 95 | ||
DV585005.1 | - | 2 - 3 | 269 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
