Assay Details
Gene Symbol: rtkn2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005937.2 | NP_001005937.2 | 8 - 9 | 1004 | 59 | |
GenBank mRNA | BC083519.1 | - | 8 - 9 | 1004 | 59 | |
BC092947.1 | - | 8 - 9 | 1005 | 59 | ||
BC098873.1 | - | 8 - 9 | 1004 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
