Assay Details
Gene Symbol: mettl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005949.1 | NP_001005949.1 | 3 - 4 | 456 | 99 | |
GenBank mRNA | BC083455.1 | - | 3 - 4 | 456 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
