Assay Details
Gene Symbol: arl6ip5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006096.1 | NP_001006096.1 | 1 - 2 | 243 | 100 | |
GenBank mRNA | BC083221.1 | - | 1 - 2 | 243 | 100 | |
BC152156.1 | - | 1 - 2 | 430 | 100 | ||
BC164985.1 | - | 1 - 2 | 209 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
