Assay Details
Gene Symbol: arl6ip5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006096.1 | NP_001006096.1 | 2 - 3 | 462 | 76 | |
GenBank mRNA | BC083221.1 | - | 2 - 3 | 462 | 76 | |
BC164985.1 | - | 2 - 3 | 428 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
