Assay Details
Gene Symbol: hsd17b10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006098.1 | NP_001006098.1 | 2 - 3 | 256 | 113 | |
GenBank mRNA | BC083219.1 | - | 2 - 3 | 256 | 113 | |
BC165246.1 | - | 2 - 3 | 215 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
