Assay Details
Gene Symbol: btg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007351.2 | NP_001007352.1 | 2 - 3 | 319 | 60 | |
NM_001326706.1 | NP_001313635.1 | 3 - 4 | 395 | 60 | ||
NM_001326707.1 | NP_001313636.1 | 2 - 3 | 336 | 60 | ||
NM_001326708.1 | NP_001313637.1 | 2 - 3 | 319 | 60 | ||
GenBank mRNA | BC085526.1 | - | 2 - 3 | 269 | 60 | |
BC164876.1 | - | 1 - 2 | 206 | 60 | ||
EH539756.1 | - | 2 - 3 | 298 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
