Assay Details
Gene Symbol: pus1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008603.2 | NP_001008603.2 | 2 - 3 | 472 | 97 | |
GenBank mRNA | BC086827.1 | - | 2 - 3 | 377 | 97 | |
EH545031.1 | - | 2 - 3 | 479 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
