Assay Details
Gene Symbol: ndufaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013308.2 | NP_001013326.1 | 5 - 6 | 1086 | 127 | |
XM_005156828.4 | XP_005156885.1 | 3 - 4 | 934 | 127 | ||
XM_005156829.4 | XP_005156886.1 | 5 - 6 | 1112 | 127 | ||
XM_005156830.4 | XP_005156887.1 | 4 - 5 | 1050 | 127 | ||
XM_005156831.4 | XP_005156888.1 | 4 - 5 | 1053 | 127 | ||
XM_005156832.4 | XP_005156889.1 | 3 - 4 | 987 | 127 | ||
XM_009307315.3 | XP_009305590.1 | 5 - 6 | 1160 | 127 | ||
XM_017358738.2 | XP_017214227.1 | 4 - 5 | 1107 | 127 | ||
GenBank mRNA | BC090683.1 | - | 5 - 6 | 1090 | 127 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
