Assay Details
Gene Symbol: aspscr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017575.1 | NP_001017575.1 | 2 - 3 | 170 | 100 | |
GenBank mRNA | BC092736.1 | - | 2 - 3 | 170 | 100 | |
BC164415.1 | - | 2 - 3 | 175 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
