Assay Details
Gene Symbol: rgs16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017846.1 | NP_001017846.1 | 2 - 3 | 192 | 88 | |
GenBank mRNA | BC092767.1 | - | 2 - 3 | 192 | 88 | |
BC164299.1 | - | 2 - 3 | 187 | 88 | ||
BC171682.1 | - | 2 - 3 | 178 | 88 | ||
BC171686.1 | - | 2 - 3 | 178 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
