Assay Details
Gene Symbol: acot9.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020744.1 | NP_001018580.1 | 13 - 14 | 1152 | 64 | |
XM_005162711.4 | XP_005162768.1 | 14 - 15 | 1222 | 64 | ||
GenBank mRNA | BC095220.1 | - | 13 - 14 | 1152 | 64 | |
BC159215.1 | - | 14 - 15 | 1115 | 64 | ||
BC171360.1 | - | 14 - 15 | 1170 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
