Assay Details
Gene Symbol: acot9.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020744.1 | NP_001018580.1 | 2 - 3 | 198 | 73 | |
GenBank mRNA | BC095220.1 | - | 2 - 3 | 198 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
