Assay Details
Gene Symbol: arl6ip4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020758.1 | NP_001018594.1 | 4 - 5 | 650 | 80 | |
GenBank mRNA | BC081670.1 | - | 4 - 5 | 673 | 80 | |
BC095624.1 | - | 4 - 5 | 650 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
