Assay Details
Gene Symbol: mrpl30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020761.2 | NP_001018597.1 | 3 - 4 | 511 | 146 | |
GenBank mRNA | BC095668.1 | - | 3 - 4 | 513 | 146 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
