Assay Details
Gene Symbol: prdx5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024406.1 | NP_001019577.1 | 3 - 4 | 296 | 104 | |
GenBank mRNA | BC095755.1 | - | 3 - 4 | 296 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
