Assay Details
Gene Symbol: nsun3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025302.1 | NP_001020473.1 | 2 - 3 | 281 | 75 | |
XM_005159919.4 | XP_005159976.1 | 2 - 3 | 381 | 75 | ||
GenBank mRNA | BC076487.1 | - | 2 - 3 | 233 | 75 | |
BC098531.1 | - | 2 - 3 | 281 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
