Assay Details
Gene Symbol: ube2s | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025536.1 | NP_001020707.1 | 2 - 3 | 635 | 60 | |
GenBank mRNA | BC097122.1 | - | 2 - 3 | 635 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
